Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Lys(Fmoc)-OH 95%

Supplier: AMBEED, INC
Warning

A335064-100G A335064-10G A335064-25G A335064-500G
77290-472EA 83 USD InStock
77290-472 77290-474 77290-476 77290-478
Fmoc-Lys(Fmoc)-OH 95%
Fmoc-Lys(Fmoc)-OH
Precaución: for Research Use Only


Formula: C₃₆H₃₄N₂O₆
MW: 590.676 g/mol
MDL Number: MFCD00080275
CAS Number: 78081-87-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty