Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethoxy)salicylaldehyde 95%

Supplier: AMBEED, INC
Warning

A336387-10G A336387-1G A336387-25G A336387-5G
77239-818EA 76 USD InStock
77239-818 77239-820 77239-822 77239-824
5-(Trifluoromethoxy)salicylaldehyde 95%
5-(Trifluoromethoxy)salicylaldehyde
Precaución: for Research Use Only


Formula: C₈H₅F₃O₃
MDL Number: MFCD00075249
CAS Number: 93249-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty