Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Thiophenedicarboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A337154-10G A337154-1G A337154-250MG A337154-5G
77251-352EA 161 USD InStock
77251-352 77251-354 77251-356 77251-358
2,3-Thiophenedicarboxaldehyde 98%
2,3-Thiophenedicarboxaldehyde
Precaución: for Research Use Only


Formula: C₆H₄O₂S
MW: 140.16 g/mol
MDL Number: MFCD00209616
CAS Number: 932-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty