Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydrozoline hydrochloride 98%

Supplier: AMBEED, INC
Warning

A339557-1G A339557-1MG A339557-25G A339557-5G
77277-932EA 14 USD InStock
77277-932 77277-934 77277-936 77277-938
Tetrahydrozoline hydrochloride 98%
Tetrahydrozoline hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₇ClN₂
MDL Number: MFCD00058029
CAS Number: 522-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty