Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-yn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide 98%

Supplier: AMBEED, INC
Warning

A342706-10MG A342706-1MG A342706-50MG A342706-5MG
77239-904EA 85 USD InStock
77239-904 77239-906 77239-908 77239-910
(R)-2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-yn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide 98%
(R)-2-Amino-1-ethyl-7-(3-hydroxy-4-methoxy-3-methylbut-1-yn-1-yl)-N-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₈H₂₂N₄O₄
MDL Number: MFCD23098773
CAS Number: 1433953-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty