Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoroanisole 95%

Supplier: AMBEED, INC
Warning

A343266-100G A343266-10G A343266-1G A343266-25G A343266-5G
77278-008EA 528 USD InStock
77278-008 77278-010 77278-012 77278-014 77278-016
3-Bromo-2-fluoroanisole 95%
3-Bromo-2-fluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrFO
MDL Number: MFCD07780183
CAS Number: 295376-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty