Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Chloro-α,α,α-trifluoro-o-xylene 95%

Supplier: AMBEED, INC
Danger

A344201-10G A344201-1G A344201-25G A344201-5G
77251-440EA 25 USD InStock
77251-440 77251-442 77251-444 77251-446
α'-Chloro-α,α,α-trifluoro-o-xylene 95%
α'-Chloro-α,α,α-trifluoro-o-xylene
Precaución: for Research Use Only


Formula: C₈H₆ClF₃
MW: 194.58 g/mol
MDL Number: MFCD00009920
CAS Number: 21742-00-7
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty