Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluorobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A344395-100G A344395-10G A344395-1G A344395-25G A344395-5G
77290-572EA 89 USD InStock
77290-572 77290-574 77290-576 77290-578 77290-580
4-Chloro-2-fluorobenzyl alcohol 98%
4-Chloro-2-fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆ClFO
MDL Number: MFCD00143285
CAS Number: 56456-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty