Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

15-(tert-Butoxy)-15-oxopentadecanoic acid 99%

Supplier: AMBEED, INC
Warning

A346074-100G A346074-1G A346074-25G A346074-5G
77263-810EA 386 USD InStock
77263-810 77263-812 77263-814 77263-816
15-(tert-Butoxy)-15-oxopentadecanoic acid 99%
15-(tert-Butoxy)-15-oxopentadecanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₃₆O₄
MW: 328.48 g/mol
MDL Number: MFCD30470973
CAS Number: 905302-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty