Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethyl-2-hydroxypyrimidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A346138-100G A346138-10G A346138-25G A346138-5G
77290-598EA 57 USD InStock
77290-598 77290-600 77290-602 77290-604
4,6-Dimethyl-2-hydroxypyrimidine hydrochloride 98%
4,6-Dimethyl-2-hydroxypyrimidine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₈N₂O
MDL Number: MFCD00035312
CAS Number: 34289-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty