Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(N-(butylcarbamoyl)sulfamoyl)benzoate 97%

Supplier: AMBEED, INC
Warning

A346337-100MG A346337-1G A346337-250MG
77251-482EA 75 USD InStock
77251-482 77251-484 77251-486
Methyl 4-(N-(butylcarbamoyl)sulfamoyl)benzoate 97%
Methyl 4-(N-(butylcarbamoyl)sulfamoyl)benzoate


Formula: C₁₃H₁₈N₂O₅S
MDL Number: MFCD00143931
CAS Number: 88241-94-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty