Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-nitro-4-(trifluoromethyl)benzene 98%

Supplier: AMBEED, INC
Warning

A346759-1G A346759-250MG A346759-25G
77290-606EA 28 USD InStock
77290-606 77290-608 77290-610
1-Iodo-2-nitro-4-(trifluoromethyl)benzene 98%
1-Iodo-2-nitro-4-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₇H₃F₃INO₂
MDL Number: MFCD07783651
CAS Number: 400-97-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty