Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A346873-100G A346873-10G A346873-1G A346873-250MG A346873-25G A346873-5G
77290-612EA 195 USD InStock
77290-612 77290-614 77290-616 77290-618 77290-620 77290-622
4-Bromo-3-methoxybenzoic acid 95%
4-Bromo-3-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD08436017
CAS Number: 56256-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty