Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Bromomethyl)naphthalene 98%

Supplier: AMBEED, INC
Danger

A348707-100G A348707-10G A348707-1G A348707-25G A348707-500G A348707-5G
77239-968EA 96 USD InStock
77239-968 77239-970 77239-972 77239-974 77239-976 77239-978
2-(Bromomethyl)naphthalene 98%
2-(Bromomethyl)naphthalene
Precaución: for Research Use Only


Formula: C₁₁H₉Br
MDL Number: MFCD00004123
CAS Number: 939-26-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty