Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apto-253 98%

Supplier: AMBEED, INC
Warning

A350879-10MG A350879-1MG A350879-25MG A350879-50MG A350879-5MG
77251-538EA 120 USD InStock
77251-538 77251-540 77251-542 77251-544 77251-546
Apto-253 98%
Apto-253
Precaución: for Research Use Only


Formula: C₂₂H₁₄FN₅
MDL Number: MFCD25976826
CAS Number: 916151-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty