Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chlorooxindole 97%

Supplier: AMBEED, INC
Warning

A353970-100G A353970-1G A353970-25G A353970-5G
77278-200EA 167 USD InStock
77278-200 77278-202 77278-204 77278-206
5-Chlorooxindole 97%
5-Chlorooxindole
Precaución: for Research Use Only


Formula: C₈H₆ClNO
MDL Number: MFCD00191927
CAS Number: 17630-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty