Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,11-Dimethyl-6H-pyrido[4,3-b]carbazole hydrochloride 98%

Supplier: AMBEED, INC
Warning

A354526-1MG A354526-50MG A354526-5MG
77263-908EA 30 USD InStock
77263-908 77263-910 77263-912
5,11-Dimethyl-6H-pyrido[4,3-b]carbazole hydrochloride 98%
5,11-Dimethyl-6H-pyrido[4,3-b]carbazole hydrochloride
Precaución: for Research Use Only


Formula: C₁₇H₁₅ClN₂
MDL Number: MFCD00050600
CAS Number: 5081-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty