Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-2-methylquinolin-4(1H)-one 97%

Supplier: AMBEED, INC
Warning

A355195-100MG A355195-1G A355195-250MG A355195-5G
77240-046EA 55 USD InStock
77240-046 77240-048 77240-050 77240-052
1-Ethyl-2-methylquinolin-4(1H)-one 97%
1-Ethyl-2-methylquinolin-4(1H)-one
Precaución: for Research Use Only


Formula: C₁₂H₁₃NO
MDL Number: MFCD20534753
CAS Number: 15574-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty