Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylethyl phenyl ether 97%

Supplier: AMBEED, INC
Warning

A355453-10G A355453-1G A355453-5G
77278-210EA 54 USD InStock
77278-210 77278-212 77278-214
2-Phenylethyl phenyl ether 97%
2-Phenylethyl phenyl ether
Precaución: for Research Use Only


Formula: C₁₄H₁₄O
MDL Number: MFCD00043544
CAS Number: 40515-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty