Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylphosphonium tetrafluoroborate 98%

Supplier: AMBEED, INC
Warning

A356704-250MG A356704-5G A356704-100MG
77251-614EA 42 USD InStock
77251-614 77251-616 77746-700
Triisopropylphosphonium tetrafluoroborate 98%
Triisopropylphosphonium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₉H₂₂BF₄P
MDL Number: MFCD06796644
CAS Number: 121099-07-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty