Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(p-cymene)triphenylphosphineruthenium(II) 98%

Supplier: AMBEED, INC
Warning

A357714-1G A357714-250MG A357714-5G
77251-628EA 14.42 USD InStock
77251-628 77251-630 77251-632 77251-634
Dichloro(p-cymene)triphenylphosphineruthenium(II) 98%
Dichloro(p-cymene)triphenylphosphineruthenium(II)
Precaución: for Research Use Only


Formula: C₂₈H₂₉Cl₂PRu
MW: 568.48 g/mol
MDL Number: MFCD28144560
CAS Number: 52490-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty