Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-Me-Gln(Trt)-OH 97%

Supplier: AMBEED, INC
Warning

A360020-100MG A360020-250MG A360020-1G
77240-110EA 9 USD InStock
77240-110 77240-112 77746-713
Fmoc-N-Me-Gln(Trt)-OH 97%
Fmoc-N-Me-Gln(Trt)-OH
Precaución: for Research Use Only


Formula: C₄₀H₃₆N₂O₅
MW: 624.72 g/mol
MDL Number: MFCD22200774
CAS Number: 1632075-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty