Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isopropylnicotinic acid 97%

Supplier: AMBEED, INC
Warning

A363504-100MG A363504-1G A363504-250MG A363504-5G
77290-796EA 40 USD InStock
77290-796 77290-798 77290-800 77290-802
5-Isopropylnicotinic acid 97%
5-Isopropylnicotinic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.18 g/mol
MDL Number: MFCD11043004
CAS Number: 73591-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty