Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-3-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A363678-10G A363678-1G A363678-250MG A363678-5G
77278-358EA 75 USD InStock
77278-358 77278-360 77278-362 77278-364
5-Methoxy-3-pyridineboronic acid pinacol ester 97%
5-Methoxy-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₈BNO₃
MW: 235.0872 g/mol
MDL Number: MFCD03412068
CAS Number: 445264-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty