Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

APS-2-79 hydrochloride 98%

Supplier: AMBEED, INC
Danger

A364339-10MG A364339-1MG A364339-25MG A364339-50MG A364339-5MG
77264-036EA 20 USD InStock
77264-036 77264-038 77264-040 77264-042 77264-044
APS-2-79 hydrochloride 98%
APS-2-79 hydrochloride
Precaución: for Research Use Only


Formula: C₂₃H₂₂ClN₃O₃
MDL Number: MFCD30536203
CAS Number: 2002381-31-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty