Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isopropyl 4-boronobenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A365048-1G A365048-250MG A365048-5G
77290-822EA 139 USD InStock
77290-822 77290-824 77290-826
N-Isopropyl 4-boronobenzenesulphonamide 98%
N-Isopropyl 4-boronobenzenesulphonamide
Precaución: for Research Use Only


Formula: C₉H₁₄BNO₄S
MW: 243.08 g/mol
MDL Number: MFCD07363752
CAS Number: 850589-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty