Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,4-dichlorobenzoate 98%

Supplier: AMBEED, INC
Warning

A365887-100G A365887-1G A365887-25G A365887-5G
77240-172EA 110 USD InStock
77240-172 77240-174 77240-176 77240-178
Methyl-2,4-dichlorobenzoate 98%
Methyl-2,4-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₂
MDL Number: MFCD00085117
CAS Number: 35112-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty