Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,3-Trimethyl-3-phenylindan 98%

Supplier: AMBEED, INC
Warning

A366198-10G A366198-1G A366198-25G A366198-5G
77240-180EA 219 USD InStock
77240-180 77240-182 77240-184 77240-186
1,1,3-Trimethyl-3-phenylindan 98%
1,1,3-Trimethyl-3-phenylindan
Precaución: for Research Use Only


Formula: C₁₈H₂₀
MW: 236.36
Boiling Pt: 308.5
Melting Pt: 52.5
Density: 1.0009
MDL Number: MFCD00021240
CAS Number: 3910-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty