Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobenzene-1,4-diamine 95%

Supplier: AMBEED, INC
Warning

A367182-100MG A367182-10G A367182-1G A367182-250MG A367182-25G A367182-5G
77264-068EA 14 USD InStock
77264-068 77264-070 77264-072 77264-074 77264-076 77264-078
2-Bromobenzene-1,4-diamine 95%
2-Bromobenzene-1,4-diamine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂
MDL Number: MFCD06411033
CAS Number: 13296-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty