Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Allyloxy-2-hydroxypropane sulfonate sodium salt Active Material 40%

Supplier: AMBEED, INC
Warning

A367720-10G A367720-25G A367720-5G
77278-400EA 10 USD InStock
77278-400 77278-402 77278-404
3-Allyloxy-2-hydroxypropane sulfonate sodium salt Active Material 40%
3-Allyloxy-2-hydroxypropane sulfonate sodium salt
Precaución: for Research Use Only


Formula: C₆H₁₁NaO₅S
MW: 218.199 g/mol
MDL Number: MFCD00080396
CAS Number: 52556-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty