Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-1439 98%

Supplier: AMBEED, INC
Warning

A367825-100MG A367825-50MG
77251-730EA 407.99 USD InStock
77251-730 77251-732
MK-1439 98%
MK-1439
Precaución: for Research Use Only


Formula: C₁₇H₁₁ClF₃N₅O₃
MW: 425.75 g/mol
MDL Number: MFCD22417167
CAS Number: 1338225-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty