Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Propane-2,2-diyl)bis(2-allylphenol) 90%

Supplier: AMBEED, INC
Warning

A367951-100G A367951-25G A367951-500G A367951-1000G
77251-734EA 13 USD InStock
77251-734 77251-736 77251-738 77551-066
4,4'-(Propane-2,2-diyl)bis(2-allylphenol) 90%
4,4'-(Propane-2,2-diyl)bis(2-allylphenol)
Precaución: for Research Use Only


Formula: C₂₁H₂₄O₂
MDL Number: MFCD00191977
CAS Number: 1745-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty