Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A368126-10G A368126-25G
77290-864EA 7 USD InStock
77290-864 77290-866
Sodium-4-hydroxybenzoate 98%
Sodium-4-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₇H₆O₃
MW: 160.10 g/mol
MDL Number: MFCD00016530
CAS Number: 114-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty