Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3,3-diphenylpropane 98%

Supplier: AMBEED, INC
Danger

A370084-1G A370084-5G
77278-416EA 48 USD InStock
77278-416 77278-418
1-Bromo-3,3-diphenylpropane 98%
1-Bromo-3,3-diphenylpropane
Precaución: for Research Use Only


Formula: (C₆H₅)₂CHCH₂CH₂Br
MDL Number: MFCD00044829
CAS Number: 20017-68-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty