Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Hydroxymethyl)-2-phenylacetamide 97%

Supplier: AMBEED, INC
Warning

A372445-10G A372445-250MG A372445-5G A372445-1G
77240-272EA 251.01 USD InStock
77240-272 77240-274 77240-276 77746-750
N-(Hydroxymethyl)-2-phenylacetamide 97%
N-(Hydroxymethyl)-2-phenylacetamide
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MDL Number: MFCD00021962
CAS Number: 6291-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty