Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate 97%

Supplier: AMBEED, INC
Danger

A373813-10G A373813-1G A373813-5G
77251-800EA 50 USD InStock
77251-800 77251-802 77251-804
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate 97%
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₄O₃S
MW: 286.25 g/mol
MDL Number: MFCD00155960
CAS Number: 786-31-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty