Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3',5,5'-Tetramethylbiphenyl-4,4'-diol 98%

Supplier: AMBEED, INC
Warning

A377064-100G A377064-10G A377064-25G A377064-500G A377064-5G
77290-924EA 50 USD InStock
77290-924 77290-926 77290-928 77290-930 77290-932 77290-934
3,3',5,5'-Tetramethylbiphenyl-4,4'-diol 98%
3,3',5,5'-Tetramethylbiphenyl-4,4'-diol
Precaución: for Research Use Only


Formula: C₁₆H₁₈O₂
MDL Number: MFCD00094737
CAS Number: 2417-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty