Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethylquinaldinium iodide 98%

Supplier: AMBEED, INC
Warning

A377409-100G A377409-1G A377409-25G A377409-5G
77240-328EA 156 USD InStock
77240-328 77240-330 77240-332 77240-334
1-Ethylquinaldinium iodide 98%
1-Ethylquinaldinium iodide
Precaución: for Research Use Only


Formula: C₁₂H₁₄IN
MW: 299.15 g/mol
Melting Pt: 234
MDL Number: MFCD00031829
CAS Number: 606-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty