Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((3-Boronophenyl)amino)-4-oxobutanoic acid 98%

Supplier: AMBEED, INC
Warning

A377562-1G A377562-250MG A377562-5G
77278-498EA 64 USD InStock
77278-498 77278-500 77278-502
4-((3-Boronophenyl)amino)-4-oxobutanoic acid 98%
4-((3-Boronophenyl)amino)-4-oxobutanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂BNO₅
MDL Number: MFCD01683353
CAS Number: 31754-00-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty