Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vidofludimus 98%

Supplier: AMBEED, INC
Warning

A377932-10MG A377932-1MG
77240-336EA 105 USD InStock
77240-336 77240-338
Vidofludimus 98%
Vidofludimus
Precaución: for Research Use Only


Formula: C₂₀H₁₈FNO₄
MW: 355.36 g/mol
MDL Number: MFCD18633262
CAS Number: 717824-30-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty