Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Diethylamino)-2-propanol 97%

Supplier: AMBEED, INC
Danger

A378775-25G A378775-5G
77240-348EA 41 USD InStock
77240-348 77240-350
1-(Diethylamino)-2-propanol 97%
1-(Diethylamino)-2-propanol
Precaución: for Research Use Only


Formula: C₇H₁₇NO
MDL Number: MFCD00004535
CAS Number: 4402-32-8
UN: 2734
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty