Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',4',6'-Trihydroxypropiophenone 95%

Supplier: AMBEED, INC
Warning

A379901-100MG A379901-1G A379901-1MG A379901-250MG A379901-25MG A379901-5G
77251-862EA 14 USD InStock
77251-862 77251-864 77251-866 77251-868 77251-870 77251-872
2',4',6'-Trihydroxypropiophenone 95%
2',4',6'-Trihydroxypropiophenone
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MDL Number: MFCD00016456
CAS Number: 2295-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty