Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-PEG3-acid 97%

Supplier: AMBEED, INC
Warning

A380179-100MG A380179-250MG A380179-5G
77240-366EA 18 USD InStock
77240-366 77240-368 77240-370
m-PEG3-acid 97%
m-PEG3-acid
Precaución: for Research Use Only


Formula: C₈H₁₆O₅
MDL Number: MFCD20646047
CAS Number: 209542-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty