Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK1325756 98+%

Supplier: AMBEED, INC
Warning

A380808-10MG A380808-1MG A380808-5MG
77251-874EA 86 USD InStock
77251-874 77251-876 77251-878
GSK1325756 98+%
GSK1325756
Precaución: for Research Use Only


Formula: C₁₉H₂₁ClFN₃O₄S
MDL Number: MFCD27987922
CAS Number: 954126-98-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty