Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylhexyl bromide 99% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with potassium carbonate 1%

A381198-100G A381198-25G A381198-500G A381198-1000G
77264-242EA 18 USD InStock
77264-242 77264-244 77264-246 77748-768
2-Ethylhexyl bromide 99% stabilized
2-Ethylhexyl bromide
Precaución: for Research Use Only


Formula: C₈H₁₇Br
MDL Number: MFCD00000220
CAS Number: 18908-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty