Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-methylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A381794-10G A381794-1G A381794-250MG A381794-25G A381794-5G
77251-890EA 103 USD InStock
77251-890 77251-892 77251-894 77251-896 77251-898
2-Amino-6-methylbenzonitrile 97%
2-Amino-6-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂
MDL Number: MFCD00058686
CAS Number: 56043-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty