Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4''-Tris(diphenylamino)triphenylamine ≥95%

Supplier: AMBEED, INC
Warning

A383997-100MG A383997-250MG
77291-010EA 32 USD InStock
77291-010 77291-012
4,4',4''-Tris(diphenylamino)triphenylamine ≥95%
4,4',4''-Tris(diphenylamino)triphenylamine
Precaución: for Research Use Only


Formula: C₅₄H₄₂N₄
MDL Number: MFCD01310677
CAS Number: 105389-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty