Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylcaproic acid 98%

Supplier: AMBEED, INC
Danger

A385849-100G A385849-25G A385849-500G
77264-302EA 13 USD InStock
77264-302 77264-304 77264-306
2-Ethylcaproic acid 98%
2-Ethylcaproic acid
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MDL Number: MFCD00002675
CAS Number: 149-57-5
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty