Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-indole 98%

Supplier: AMBEED, INC
Warning

A388760-1G A388760-25G A388760-5G
77240-490EA 5 USD InStock
77240-490 77240-492 77240-494
1-Boc-indole 98%
1-Boc-indole
Precaución: for Research Use Only


Formula: C₁₃H₁₅NO₂
MW: 217.27 g/mol
MDL Number: MFCD02093939
CAS Number: 75400-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty