Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Leniolisib 98%

Supplier: AMBEED, INC
Warning

A389627-1MG A389627-25MG A389627-5MG
77264-378EA 17 USD InStock
77264-378 77264-380 77264-382
Leniolisib 98%
Leniolisib
Precaución: for Research Use Only


Formula: C₂₁H₂₅F₃N₆O₂₅
MDL Number: MFCD30470232
CAS Number: 1354690-24-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty